Thermo Scientific Chemicals

Diaion™ SK1B(Na), ion exchange resin, 8% cross-linked, strongly acidic gel type, 2.0 meq/ml on PS-DVB, Thermo Scientific Chemicals

Catalog number: 046398.A1
1 kg, Each
Thermo Scientific Chemicals

Diaion™ SK1B(Na), ion exchange resin, 8% cross-linked, strongly acidic gel type, 2.0 meq/ml on PS-DVB, Thermo Scientific Chemicals

Catalog number: 046398.A1
1 kg, Each
Quantity
Catalog number: 046398.A1
also known as 046398-A1
Price (USD)
Quantity
-
Have Questions?
Assay from Supplier's CofA
Water Content 43-50%
Exchange capacity
≥2.0 meq/ml
Form
Beads
Appearance (Color)
Brown
Particle Size
300-1180?m

Description

Water softening, metal recovery, dehydration of solvents, amino acid purification

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text