Thermo Scientific Chemicals

Methoxypolyethylene glycol amine, M.W. 10,000, Thermo Scientific Chemicals

Catalog number: 046433.03
1 g, Each
Thermo Scientific Chemicals

Methoxypolyethylene glycol amine, M.W. 10,000, Thermo Scientific Chemicals

Catalog number: 046433.03
1 g, Each
Quantity
Catalog number: 046433.03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
80506-64-5
Molecular Formula
H2NCH2CH2(OCH2CH2)nOCH3
InChI Key
QWCGXANSAOXRFE-UHFFFAOYSA-N
SMILES
COCCOCCN
MDL Number
MFCD00131599
Form
powder
Proton NMR
> 95%
Appearance (Color)
Off-white to pale yellow

Description

It is used as a reagent for pegylation of biomolecules, improvement of solubility and other pharmacological parameters, for the preparation of various conjugates containing polyethylene glycol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text