Thermo Scientific Chemicals
Palladium, 1% on granular carbon, reduced, Thermo Scientific Chemicals
Catalog number: 046462.36
500 g, Each
Thermo Scientific Chemicals
Palladium, 1% on granular carbon, reduced, Thermo Scientific Chemicals
Catalog number: 046462.36
500 g, Each
Quantity
Catalog number: 046462.36
also known as 046462-36
Price (USD)
Quantity
-
Have Questions?
Description
It is used in hydrogenation of aromatic and aliphatic nitro groups; in reductive alkylation/amination; hydrogenation of aromatic carbonyls, olefins, nitriles and as a catalyst in the Suzuki reaction and Stille reaction.