Thermo Scientific Chemicals

Polyethylene glycol diacrylate, M.W. 3,400, Thermo Scientific Chemicals

Catalog number: 046497.03
1 g, Each
Thermo Scientific Chemicals

Polyethylene glycol diacrylate, M.W. 3,400, Thermo Scientific Chemicals

Catalog number: 046497.03
1 g, Each
Quantity
Catalog number: 046497.03
also known as 046497-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
26570-48-9
IUPAC Name
ethane-1,2-diol; prop-2-enoic acid
Molecular Formula
C5H10O4
InChI Key
FCDZZFLRLVQGEH-UHFFFAOYSA-N
SMILES
OCCO.OC(=O)C=C
Assay by NMR
> 95%
Comment
Inhibited with a minimum of 300 ppm MEHQ
Appearance (Color)
Off-white to light yellow
Assay (unspecified)
Mn ~ 3000
Form
Solid

Description

Polyethylene glycol diacrylate is used in dental compounds, offset inks, varnishes and flexible coatings. It widely used in various biomedical applications such as hydrogel development for drug delivery, tissue engineering, wound healing and electrospinning processes for bioapplication due to its non-toxicity and cytocompatiblity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text