Chemical Identifiers
CAS165324-09-4
IUPAC Namecalcium bis(trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane)
Molecular FormulaC4CaF12N2O8S4
InChI KeyWFABOCFDABTAPE-UHFFFAOYSA-N
SMILES[Ca++].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
View more