Thermo Scientific Chemicals

Calcium bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals

Catalog number: 046533.06
5 g, Each
Thermo Scientific Chemicals

Calcium bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals

Catalog number: 046533.06
5 g, Each
Quantity
Catalog number: 046533.06
also known as 046533-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
165324-09-4
IUPAC Name
calcium bis(trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane)
Molecular Formula
C4CaF12N2O8S4
InChI Key
WFABOCFDABTAPE-UHFFFAOYSA-N
SMILES
[Ca++].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Appearance (Color)
White
Form
Granular solid
Fluorine-19 NMR
Report value

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text