Thermo Scientific Chemicals

Polyethylene glycol methyl ether acrylate, M.W. 1,000, Thermo Scientific Chemicals

Catalog number: 046537.03
1 g, Each
Thermo Scientific Chemicals

Polyethylene glycol methyl ether acrylate, M.W. 1,000, Thermo Scientific Chemicals

Catalog number: 046537.03
1 g, Each
Quantity
Catalog number: 046537.03
also known as 046537-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
32171-39-4
Molecular Formula
(C2H4O)nC4H6O2
InChI Key
HFCUBKYHMMPGBY-UHFFFAOYSA-N
SMILES
COCCOC(=O)C=C
MDL Number
MFCD00803685
Appearance (Color)
Yellow
Form
Viscous liquid
Proton NMR
Conforms to structure

Description

Polyethylene glycol methyl ether acrylate is used as a raw material in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text