Thermo Scientific Chemicals

Sodium methoxide, ca 25% w/w in methanol, Thermo Scientific Chemicals

Catalog number: 046585.AP
500 mL, Each
Thermo Scientific Chemicals

Sodium methoxide, ca 25% w/w in methanol, Thermo Scientific Chemicals

Catalog number: 046585.AP
500 mL, Each
Quantity
Catalog number: 046585.AP
also known as 046585-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
124-41-4
IUPAC Name
sodium methanolate
Molecular Formula
CH3NaO
InChI Key
WQDUMFSSJAZKTM-UHFFFAOYSA-N
SMILES
[Na+].C[O-]
Appearance (Color)
Colorless to yellow
Assay (Titration)
24.4 to 26.0 wt%
Chloride content
10 ppm, max
Water Content (Karl Fischer Titration)
0.1% max
Form
liquid

Description

Sodium methoxide 2% w/w in methanol is used in the preparation of high-spin hexanuclear Mn(III) cluster, [NaMn6 (OMe)12 (dbm)6]+. It is useful in the synthesis of pharmaceuticals and agrochemicals. It serves as an initiator of anionic addition polymerization with ethylene oxide, forming a polyether with high molecular weight.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text