Chloro(crotyl)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II), Thermo Scientific Chemicals
Chloro(crotyl)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chloro(crotyl)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II), Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 046639.03
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1252598-33-6
Molecular FormulaC52H54ClFePPd
SMILESCl[Pd++].C\C=C/C.CC(C)(C)P(c1cccc1[Fe])C(C)(C)C.C1=CC=C(C=C1)c1c(c(c(c1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)907.69
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)5.60-6.60% Hydrogen
Assay (unspecified)67.80-69.80% Carbon
Proton NMRPass
Carbon-13 NMRPass
Chloro(crotyl)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II) acts as a ligand and chiral catalyst in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only