Thermo Scientific Chemicals

Sodium bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals

Catalog number: 046640.MD
250 mg, Each
Thermo Scientific Chemicals

Sodium bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals

Catalog number: 046640.MD
250 mg, Each
Quantity
Catalog number: 046640.MD
also known as 046640-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
91742-21-1
IUPAC Name
1,1,1-trifluoro-N-trifluoromethanesulfonylmethanesulfonamide sodium
Molecular Formula
C2HF6NNaO4S2
InChI Key
QXZNUMVOKMLCEX-UHFFFAOYSA-N
SMILES
[Na].FC(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F
Appearance (Color)
White
Form
Powder
Assay (Titration)
≥94%

Description

Sodium bis(trifluoromethylsulfonyl)imide is used as an electrolyte in batteries and fuel cells. It also finds use as an ionic liquid and Lewis acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text