Thermo Scientific Chemicals
Zinc, Quantofix™ Test Strips
Catalog number: 046651.QA
Each
Thermo Scientific Chemicals
Zinc, Quantofix™ Test Strips
Catalog number: 046651.QA
Each
Catalog number: 046651.QA
also known as 046651-QA
Price (USD)
Quantity
-
Have Questions?
Description
Zinc, Quantofix™ Test Strips is used to measure the determination of zinc in solutions quickly. It can be used readily for measurements. It is used to regularly check the plating baths in the metal plating industry. It is also used to check the concentration of zinc in cooling water treatment, where zinc salts are used as corrosion inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Liquid
  • Colorless
  • Fully miscible