Thermo Scientific Chemicals

Dicyclopentanylmethyl acrylate, Thermo Scientific Chemicals

Catalog number: 046652.14
25 g, Each
Thermo Scientific Chemicals

Dicyclopentanylmethyl acrylate, Thermo Scientific Chemicals

Catalog number: 046652.14
25 g, Each
Quantity
Catalog number: 046652.14
also known as 046652-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
93962-84-6
IUPAC Name
{tricyclo[5.2.1.0²,⁶]decan-3-yl}methyl prop-2-enoate
Molecular Formula
C14H20O2
InChI Key
PMPNHSZLJPXGCD-UHFFFAOYNA-N
SMILES
C=CC(=O)OCC1CCC2C3CCC(C3)C12
Appearance (Color)
Pale yellow to pale brown
Viscosity
< 100 cP ( 25 deg C)
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text