Thermo Scientific™

Silicon powder, -325 mesh, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 046654.18
50 g, Each
Thermo Scientific™

Silicon powder, -325 mesh, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 046654.18
50 g, Each
Quantity
Catalog number: 046654.18
also known as 046654-18
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
7440-21-3
IUPAC Name
silicon
Molecular Formula
Si
InChI Key
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
SMILES
[Si]
Form
Powder, -325 mesh
Total Metal Impurities
≤0.001%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable solids
    Category 2
    Label Elements
    Signal Word

    Warning

    Physical Hazards

    H228 - Flammable solid

    Precautionary Statements

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking