Chemical Identifiers
CAS205319-10-4
IUPAC Name[5-(diphenylphosphanyl)-9,9-dimethyl-9H-xanthen-4-yl]diphenylphosphane; dichloropalladiumbis(ylium)
Molecular FormulaC39H32Cl2OP2Pd
InChI KeyMNBAPNBMBOYNAW-UHFFFAOYSA-L
SMILESCl[Pd++]Cl.CC1(C)C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1
View more