Thermo Scientific Chemicals

Magnesium bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals

Catalog number: 046667.06
5 g, Each
Thermo Scientific Chemicals

Magnesium bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals

Catalog number: 046667.06
5 g, Each
Quantity
Catalog number: 046667.06
also known as 046667-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
133395-16-1
IUPAC Name
magnesium(2+) bis(trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane)
Molecular Formula
C4F12MgN2O8S4
InChI Key
DMFBPGIDUUNBRU-UHFFFAOYSA-N
SMILES
[Mg++].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Form
Powder and/ or granules
Appearance (Color)
White to pale yellow
Fluorine-19 NMR
Conforms

Description

Magnesium bis(trifluoromethylsulfonyl)imide is a strong Lewis acid and used as a catalyst in organic synthesis. It is an electrolyte, which shows high anodic stability used in rechargeable magnesium batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text