Thermo Scientific Chemicals

Palladium(II) trimethylacetate, Thermo Scientific Chemicals

Catalog number: 046685.03
1 g, Each
Thermo Scientific Chemicals

Palladium(II) trimethylacetate, Thermo Scientific Chemicals

Catalog number: 046685.03
1 g, Each
Quantity
Catalog number: 046685.03
also known as 046685-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106224-36-6
IUPAC Name
palladium(2+) bis(2,2-dimethylpropanoate)
Molecular Formula
C10H18O4Pd
InChI Key
PAGZTSLSNQZYEV-UHFFFAOYSA-L
SMILES
[Pd++].CC(C)(C)C([O-])=O.CC(C)(C)C([O-])=O
Assay (unspecified)
C 37.90-39.90%

Description

Palladium(II) trimethylacetate is used as a catalyst for aerobic cyclization of acrylic acid with alkenes. It is utilized in the benzylation of heterocycles with benzyl chloride and direct arylation reactions. Furthermore, it is used in regioselective oxidative arene cross-coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text