Hastelloy C∣r gauze, 40 mesh woven from 0.19mm (0.0075in) dia wire
Hastelloy C∣r gauze, 40 mesh woven from 0.19mm (0.0075in) dia wire
Thermo Scientific Chemicals

Hastelloy C∣r gauze, 40 mesh woven from 0.19mm (0.0075in) dia wire

Have Questions?
Change viewbuttonViewtableView
Quantity:
150 x 150 mm
300 x 300 mm
Catalog number 046690.HB
also known as 046690-HB
Price (USD)
399.65
Online Exclusive
444.00
Save 44.35 (10%)
Each
In stock
Add to cart
Quantity:
300 x 300 mm
Request bulk or custom format
Price (USD)
399.65
Online Exclusive
444.00
Save 44.35 (10%)
Each
Add to cart
Specifications
Chemical Name or MaterialHastelloy C™ gauze
MDL NumberMFCD00801065
Mesh Size40 Mesh
Name Note40 mesh woven from 0.19mm (0.0075 in.) dia. wire
Recommended StorageAmbient temperatures
View more
Hastelloy C is used in combustion chambers, heat-treating equipment, collector rings, combustion chambers and exhaust stacks for high-heat applications in for conventional aircraft and jets, pipe connections, pumps and valves for oil refinery, chemical and petrochemical applications. Also used in components exposed to flue gas or flue gas desulfurization plants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection