Tantalum gauze, 100 mesh woven from 0.08mm (0.003 in.) dia. wire, twilled weave, Thermo Scientific Chemicals
Tantalum gauze, 100 mesh woven from 0.08mm (0.003 in.) dia. wire, twilled weave, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tantalum gauze, 100 mesh woven from 0.08mm (0.003 in.) dia. wire, twilled weave, Thermo Scientific Chemicals

Have Questions?
Quantity:
75 x 75 mm
Catalog number 046695.FL
also known as 046695-FL
Price (USD)/ Each
139.00
-
Add to cart
Quantity:
75 x 75 mm
Request bulk or custom format
Price (USD)/ Each
139.00
Add to cart
Tantalum gauze, 100 mesh woven from 0.08mm (0.003 in.) dia. wire, twilled weave, Thermo Scientific Chemicals
Catalog number046695.FL
Price (USD)/ Each
139.00
-
Add to cart
Chemical Identifiers
CAS7440-25-7
IUPAC Nametantalum
Molecular FormulaTa
InChI KeyGUVRBAGPIYLISA-UHFFFAOYSA-N
SMILES[Ta]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver
FormGauze, 100 mesh woven from 0.08 mm (0.003 in.) dia. wire, twilled weave
Assay from Supplier's CofATantalum: ≥99.9% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only