Thermo Scientific Chemicals

Diaion™ PK216(Na), ion exchange resin, 8% cross-linked, strongly acidic porous type, 1.75 meq/ml on PS-DVB, Thermo Scientific Chemicals

Catalog number: 046718.A1
1 kg, Each
Thermo Scientific Chemicals

Diaion™ PK216(Na), ion exchange resin, 8% cross-linked, strongly acidic porous type, 1.75 meq/ml on PS-DVB, Thermo Scientific Chemicals

Catalog number: 046718.A1
1 kg, Each
Quantity
Catalog number: 046718.A1
also known as 046718-A1
Price (USD)
Quantity
-
Have Questions?
Particle Size
300-1180?m
Form
Beads
Exchange capacity
≥1.75 meq/ml
Appearance (Color)
Brown
Assay from Supplier's CofA
Water Content 46-52%

Description

Used in demineralization, decolorization, metal recovery, pharmaceuticals, amino acid separation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Used in demineralization, decolorization, metal recovery, pharmaceuticals, amino acid separation
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text