Thermo Scientific Chemicals

Tetra-n-butylammonium phosphate, 98%, non-UV, Thermo Scientific Chemicals

Catalog number: 046746.04
2 g, Each
Thermo Scientific Chemicals

Tetra-n-butylammonium phosphate, 98%, non-UV, Thermo Scientific Chemicals

Catalog number: 046746.04
2 g, Each
Quantity
Catalog number: 046746.04
also known as 046746-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5574-97-0
IUPAC Name
tetrabutylazanium dihydrogen phosphate
Molecular Formula
C16H38NO4P
InChI Key
ARRNBPCNZJXHRJ-UHFFFAOYSA-M
SMILES
OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
Absorbance
at 200 nm: ≤0.20
Absorbance
at 210 nm: ≤0.15
Absorbance
at 220-400 nm: ≤0.01
Assay from Supplier's CofA
≥97.5%
Appearance (Color)
White to off-white

Description

Tetra-n-butylammonium phosphate, is used in chemicals, pharmaceutical research, pharmaceutical intermediates, and reagents. Tetrabutylammonium phosphate can be used in organic synthesis. It is used as an Ion-associating reagent for HPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text