Tetra-n-butylammonium phosphate, 98%, non-UV, Thermo Scientific Chemicals
Tetra-n-butylammonium phosphate, 98%, non-UV, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetra-n-butylammonium phosphate, 98%, non-UV, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
50 g
Catalog number 046746.09
also known as 046746-09
Price (USD)/ Each
216.65
Online exclusive
241.00 
Save 24.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
216.65
Online exclusive
241.00 
Save 24.35 (10%)
Add to cart
Tetra-n-butylammonium phosphate, 98%, non-UV, Thermo Scientific Chemicals
Catalog number046746.09
Price (USD)/ Each
216.65
Online exclusive
241.00 
Save 24.35 (10%)
-
Add to cart
Chemical Identifiers
CAS5574-97-0
IUPAC Nametetrabutylazanium dihydrogen phosphate
Molecular FormulaC16H38NO4P
InChI KeyARRNBPCNZJXHRJ-UHFFFAOYSA-M
SMILESOP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Absorbanceat 200 nm: ≤0.20
Absorbanceat 210 nm: ≤0.15
Absorbanceat 220-400 nm: ≤0.01
Assay from Supplier's CofA≥97.5%
Appearance (Color)White to off-white
View more
Tetra-n-butylammonium phosphate, is used in chemicals, pharmaceutical research, pharmaceutical intermediates, and reagents. Tetrabutylammonium phosphate can be used in organic synthesis. It is used as an Ion-associating reagent for HPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only