Palladium, 10% on activated carbon powder, Type A402023-10, nominally 50% water wet, Thermo Scientific Chemicals
Palladium, 10% on activated carbon powder, Type A402023-10, nominally 50% water wet, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Palladium, 10% on activated carbon powder, Type A402023-10, nominally 50% water wet, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
Catalog number 046789.09
also known as 046789-09
Price (USD)/ Each
-
Quantity:
10 g
Request bulk or custom format
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialPalladium
Name Note10% on activated carbon powder, Type A402023-10, Nominally 50% water wet
CAS7440-05-3
ColorBlack
Recommended StorageAmbient temperatures
View more
Palladium is used as catalyst. It is also used in jewelry, dentistry, watch making, blood sugar test strips, aircraft spark plugs and in the production of surgical instruments and electrical contacts. It is also used to make professional transverse flutes. The second-biggest application of palladium in electronics is in the manufacture of multilayer ceramic capacitors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only