Thermo Scientific Chemicals

Diaion™ SA10A(Cl), ion exchange resin, cross-linked, strongly basic gel type I, 1.3 meq/ml on PS-DVB, Thermo Scientific Chemicals

Catalog number: 046796.A1
1 kg, Each
Thermo Scientific Chemicals

Diaion™ SA10A(Cl), ion exchange resin, cross-linked, strongly basic gel type I, 1.3 meq/ml on PS-DVB, Thermo Scientific Chemicals

Catalog number: 046796.A1
1 kg, Each
Quantity
Catalog number: 046796.A1
also known as 046796-A1
Price (USD)
Quantity
-
Have Questions?
Form
Beads
Assay from Supplier's CofA
Water Content 43-47%
Particle Size
300-1180?m
Appearance (Color)
White to yellow
Exchange capacity
≥1.3 meq/ml

Description

Diaion™ SA10A(Cl) is used in the waste water treatment. It is used in demineralization, decolorization, metal recovery, high purity water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text