Polyethylene glycol diacrylate, M.W. 8,000, Thermo Scientific Chemicals
Polyethylene glycol diacrylate, M.W. 8,000, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polyethylene glycol diacrylate, M.W. 8,000, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
5 g
Catalog number 046801.03
also known as 046801-03
Price (USD)/ Each
86.65
Online exclusive
95.70 
Save 9.05 (9%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
86.65
Online exclusive
95.70 
Save 9.05 (9%)
Add to cart
Polyethylene glycol diacrylate, M.W. 8,000, Thermo Scientific Chemicals
Catalog number046801.03
Price (USD)/ Each
86.65
Online exclusive
95.70 
Save 9.05 (9%)
-
Add to cart
Chemical Identifiers
CAS26570-48-9
IUPAC Nameethane-1,2-diol; prop-2-enoic acid
Molecular FormulaC5H10O4
InChI KeyFCDZZFLRLVQGEH-UHFFFAOYSA-N
SMILESOCCO.OC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to pale cream
FormGranular solid
Assay from Supplier's CofA≥95.0%
CommentInhibitor: Report type and quantity
Molecular Weight8,000 (approx.)
Polyethylene glycol diacrylate is used to prepare photopolymerized hydrogels which are used as scaffolds for tissue regeneration and other biological applications. It acts as an important raw material for the synthesis of poly(ethylene glycol) urethane-dimethacrylates (PEGUDM) by reacting with 2-isocyanatoethyl methacrylate. It contributes to chemical resistance, flexibility, adhesion, low shrinkage, abrasion resistance and impact strength.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only