Thermo Scientific Chemicals

Sepabeads∣r SP207, synthetic adsorbent resin, highly porous type, brominated PS-DVB, P.R. 110 angstroms, Thermo Scientific Chemicals

Catalog number: 046848.A1
1 kg, Each
Thermo Scientific Chemicals

Sepabeads∣r SP207, synthetic adsorbent resin, highly porous type, brominated PS-DVB, P.R. 110 angstroms, Thermo Scientific Chemicals

Catalog number: 046848.A1
1 kg, Each
Quantity
Catalog number: 046848.A1
also known as 046848-A1
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
87924-01-4
Particle Size
250?m
Appearance (Color)
Brown
Assay from Supplier's CofA
Water Content 43-53%
Surface Area
ca. 600m²/g
Form
Beads

Description

Seabees SP207, is used in Brominating of the aromatic ring provides increased hydrophobicity, and consequently, increased selectivity for hydrophobic molecules versus conventional styrene/DVB synthetic adsorbents like Diaion™ HP20 resin. Sepabeads™ SP-207 was used in kinetic and thermodynamics investigation for adsorption/desorption behavior of anthocyanin pigments. It was used as column packing material for purification of phenolic fraction. It is widely used as a adsorbent for purification in biopharmaceutical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Bromination of the aromatic ring provides increased hydrophobicity, and consequently, increased selectivity for hydrophobic molecules versus conventional styrene/DVB synthetic adsorbents like Diaion™ HP20 resin
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text