Thermo Scientific Chemicals

Zinc bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals

Catalog number: 046880.MD
250 mg, Each
Thermo Scientific Chemicals

Zinc bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals

Catalog number: 046880.MD
250 mg, Each
Quantity
Catalog number: 046880.MD
also known as 046880-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
168106-25-0
IUPAC Name
zinc(2+) bis(trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane)
Molecular Formula
C4F12N2O8S4Zn
InChI Key
QEORIOGPVTWFMH-UHFFFAOYSA-N
SMILES
[Zn++].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Form
Powder
Appearance (Color)
White
Assay from Supplier's CofA
≥94%
Assay from Supplier's CofA
Zn: 10.1-10.7%

Description

Zinc bis(trifluoromethylsulfonyl)imide is used as a Catalyst for Enantioselective Friedel-Crafts alkylation of pyrroles with β-CF3 acrylates, Stereoselective Diels-Alder reaction, Tandem cyclopropane ring-opening / Conia-ene cyclization Aromatic nitration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text