Thermo Scientific Chemicals
Diiodo(p-cymene)ruthenium(II) dimer, Thermo Scientific Chemicals
Catalog number: 046886.06
5 g, Each
Thermo Scientific Chemicals
Diiodo(p-cymene)ruthenium(II) dimer, Thermo Scientific Chemicals
Catalog number: 046886.06
5 g, Each
Quantity
Catalog number: 046886.06
also known as 046886-06
Price (USD)
Quantity
-
Have Questions?
Description
Diiodo(p-cymene)ruthenium(II) dimer can be applied as a catalyst for many reactions such as ruthenium-catalyzed hydrogenation, preparation of Walphos-type ferrocenyl-aryl diphosphine ligands, ruthenium-catalyzed synthesis reactions, and enantioselective hydrogenation of ketones and olefins.