Thermo Scientific Chemicals

Bismuth Indium Tin ingot (Field's metal), Thermo Scientific Chemicals

Catalog number: 046895.22
100 g, Each
Thermo Scientific Chemicals

Bismuth Indium Tin ingot (Field's metal), Thermo Scientific Chemicals

Catalog number: 046895.22
100 g, Each
Quantity
Catalog number: 046895.22
also known as 046895-22
Price (USD)
Quantity
-
Have Questions?
Form
Ingot/bar
Assay from Suppliers CofA
Bismuth: 32.0-33.0%
Assay from Suppliers CofA
Indium: 50.0-52.0%
Assay from Suppliers CofA
Tin: 16.0-17.0%

Description

Bismuth Indium Tin ingot (Field's metal) is used for die casting and rapid prototyping. It acts as an alternative to Wood's metal which is an eutectic alloy of 50% bismuth, 26.7% lead, 13.3% tin, and 10% cadmium by weight.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text