Polyoxyethylene bis(amine), M.W. 8,000, Thermo Scientific Chemicals
Polyoxyethylene bis(amine), M.W. 8,000, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polyoxyethylene bis(amine), M.W. 8,000, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
5 g
Catalog number 046903.03
also known as 046903-03
Price (USD)/ Each
114.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
114.00
Add to cart
Polyoxyethylene bis(amine), M.W. 8,000, Thermo Scientific Chemicals
Catalog number046903.03
Price (USD)/ Each
114.00
-
Add to cart
Chemical Identifiers
CAS24991-53-5
SpecificationsSpecification SheetSpecification Sheet
FormGranular solid
Proton NMR≥94% ¹H nmr
Appearance (Color)White
Proton NMRConforms to structure
Molecular Weight8000
It is used as PEG modifier, its main products include high-qualified intermediates, various of amino acid and APIs. This Polymer used in the preparation of enzyme conjugates soluble in organic solvents. Promising drug carrier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only