Diaion™ WA20, ion exchange resin, weakly basic porous type, 2.5 meq/ml on poly(styrene-divinylbenzene), Thermo Scientific Chemicals
Diaion™ WA20, ion exchange resin, weakly basic porous type, 2.5 meq/ml on poly(styrene-divinylbenzene), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diaion™ WA20, ion exchange resin, weakly basic porous type, 2.5 meq/ml on poly(styrene-divinylbenzene), Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 kg
Catalog number 046918.A1
also known as 046918-A1
Price (USD)/ Each
164.65
Online exclusive
183.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
164.65
Online exclusive
183.00 
Save 18.35 (10%)
Add to cart
Diaion™ WA20, ion exchange resin, weakly basic porous type, 2.5 meq/ml on poly(styrene-divinylbenzene), Thermo Scientific Chemicals
Catalog number046918.A1
Price (USD)/ Each
164.65
Online exclusive
183.00 
Save 18.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialDiaion™ WA20, ion exchange resin
Physical Form300-1180μm Beads
Name NoteWeakly basic porous type, 2.5meq/mL on poly(styrene-divinylbenzene)
Recommended StorageAmbient temperatures
TSCANo
View more
Diaion™ WA20 is used for the removal of the strong mineral acids in standard waste water applications. They can also be applied in the special process separation such as the treatment of organic solvents. Also used in deacidification, solvent treatment, refining of gelatin, adsorption of aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only