Thermo Scientific Chemicals

Methoxypolyethylene glycol amine, M.W. 5,000, Thermo Scientific Chemicals

Catalog number: 046924.03
1 g, Each
Thermo Scientific Chemicals

Methoxypolyethylene glycol amine, M.W. 5,000, Thermo Scientific Chemicals

Catalog number: 046924.03
1 g, Each
Quantity
Catalog number: 046924.03
also known as 046924-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
80506-64-5
Molecular Formula
H2NCH2CH2(OCH2CH2)nOCH3
InChI Key
QWCGXANSAOXRFE-UHFFFAOYSA-N
SMILES
COCCOCCN
MDL Number
MFCD00131599
Form
Powder
Molecular Weight
Average Mn ~ 5000
Appearance (Color)
White to pale cream
Assay (unspecified)
> 95% (NMR)

Description

Methoxypolyethylene glycol amine is used in the reduction of immunogenicity and increasing serum half life.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text