Methoxypolyethylene glycol amine, M.W. 5,000, Thermo Scientific Chemicals
Methoxypolyethylene glycol amine, M.W. 5,000, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methoxypolyethylene glycol amine, M.W. 5,000, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 mg
1 g
Catalog number 046924.06
also known as 046924-06
Price (USD)/ Each
467.65
Online exclusive
519.00 
Save 51.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
467.65
Online exclusive
519.00 
Save 51.35 (10%)
Add to cart
Methoxypolyethylene glycol amine, M.W. 5,000, Thermo Scientific Chemicals
Catalog number046924.06
Price (USD)/ Each
467.65
Online exclusive
519.00 
Save 51.35 (10%)
-
Add to cart
Chemical Identifiers
CAS80506-64-5
Molecular FormulaH2NCH2CH2(OCH2CH2)nOCH3
InChI KeyQWCGXANSAOXRFE-UHFFFAOYSA-N
SMILESCOCCOCCN
MDL NumberMFCD00131599
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Molecular WeightAverage Mn ~ 5000
Appearance (Color)White to pale cream
Assay (unspecified)> 95% (NMR)
Methoxypolyethylene glycol amine is used in the reduction of immunogenicity and increasing serum half life.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only