Potassium peroxydisulfate, ACS, 99.0% min, Thermo Scientific Chemicals
Potassium peroxydisulfate, ACS, 99.0% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium peroxydisulfate, ACS, 99.0% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number 046939.A1
Price (USD)/ Each
153.65
Online exclusive
170.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
153.65
Online exclusive
170.00 
Save 16.35 (10%)
Add to cart
Potassium peroxydisulfate, ACS, 99.0% min, Thermo Scientific Chemicals
Catalog number046939.A1
Price (USD)/ Each
153.65
Online exclusive
170.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7727-21-1
IUPAC Namedipotassium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular FormulaK2O8S2
InChI KeyUSHAGKDGDHPEEY-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]S(=O)(=O)OOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless or white
FormSolid
Assay (unspecified)≥99.0% K2S2O8
Impurity contentChlorine compounds (as Cl): ≤0.001%
Impurity contentIron (Fe): ≤5ppm
View more
Potassium persulfate is a powerful oxidant, commonly used to initiate polymerizations. Potassium Persulphate used for bleaching and textile desizing, as an oxidizing agent and antiseptic, in purification of ammonium sulfate, and in the manufacture of soap and pharmaceuticals. It is also used as al laboratory oxidant and photography chemical. It is a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only