Thermo Scientific Chemicals

Tetrakisepoxy cyclosiloxane, Thermo Scientific Chemicals

Catalog number: 046960.22
100 g, Each
Thermo Scientific Chemicals

Tetrakisepoxy cyclosiloxane, Thermo Scientific Chemicals

Catalog number: 046960.22
100 g, Each
Quantity
Catalog number: 046960.22
also known as 046960-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
257284-60-9
IUPAC Name
2,4,6,8-tetramethyl-2,4,6,8-tetrakis({3-[(oxiran-2-yl)methoxy]propyl})-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
Molecular Formula
C28H56O12Si4
InChI Key
BLZDTVROPKIJLZ-UHFFFAOYNA-N
SMILES
C[Si]1(CCCOCC2CO2)O[Si](C)(CCCOCC2CO2)O[Si](C)(CCCOCC2CO2)O[Si](C)(CCCOCC2CO2)O1
Assay from Supplier's CofA
Epoxy Equivalent Weights (g/eq.): Report
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Colorless to pale yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text