Chloro(crotyl)(tri-tert-butylphosphine)palladium(II), Thermo Scientific Chemicals
Chloro(crotyl)(tri-tert-butylphosphine)palladium(II), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chloro(crotyl)(tri-tert-butylphosphine)palladium(II), Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
Catalog number 046962.03
also known as 046962-03
Price (USD)/ Each
370.65
Online exclusive
412.00 
Save 41.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
370.65
Online exclusive
412.00 
Save 41.35 (10%)
Add to cart
Chloro(crotyl)(tri-tert-butylphosphine)palladium(II), Thermo Scientific Chemicals
Catalog number046962.03
Price (USD)/ Each
370.65
Online exclusive
412.00 
Save 41.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1334497-00-5
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)C 47.10-49.10%
Chloro(crotyl)(tri-tert-butylphosphine)palladium(II) is also used in C-C Coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only