Platinum gauze, Unimesh N7433, expanded metal mesh, 0.34mm (0.013in) thick, Thermo Scientific Chemicals
Platinum gauze, Unimesh N7433, expanded metal mesh, 0.34mm (0.013in) thick, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum gauze, Unimesh N7433, expanded metal mesh, 0.34mm (0.013in) thick, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 046966.FF
also known as 046966-FF
Price (USD)/ Each
370.65
Online exclusive
412.00 
Save 41.35 (10%)
-
Add to cart
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)/ Each
370.65
Online exclusive
412.00 
Save 41.35 (10%)
Add to cart
Platinum gauze, Unimesh N7433, expanded metal mesh, 0.34mm (0.013in) thick, Thermo Scientific Chemicals
Catalog number046966.FF
Price (USD)/ Each
370.65
Online exclusive
412.00 
Save 41.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
FormGauze, 0.34 mm (0.013 in) thick
Total Metal ImpuritiesReport Value
Platinum gauze is used in electronics industry for electrical contacts which might be subjected to high temperatures. It also acts as a catalyst in the production of nitric acid. It plays an essential role in the cracking processes in oil refineries. It can be coated on electrode surface by electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only