Polyoxyethylene bis(amine), M.W. 3,400, Thermo Scientific Chemicals
Polyoxyethylene bis(amine), M.W. 3,400, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polyoxyethylene bis(amine), M.W. 3,400, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 mg
1 g
Catalog number 046973.06
also known as 046973-06
Price (USD)/ Each
570.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
570.00
Add to cart
Polyoxyethylene bis(amine), M.W. 3,400, Thermo Scientific Chemicals
Catalog number046973.06
Price (USD)/ Each
570.00
-
Add to cart
Specifications
Chemical Name or MaterialPolyoxyethylene bis (amine)
ColorWhite
Name NoteM.W. 3,400
CAS24991-53-5
Physical FormGranular Solid
View more
Polyoxyethylene bis(amine) with HCl, it act as reduction agent; stereo selective allylation of carbonyl compounds; in situ generation of tinenolates for directed aldol reactions. It is used in the preparation of enzyme conjugates, promising drug carrier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only