Copper foil, 0.025mm (0.001in) thick, annealed, uncoated, 99.8% (metals basis), Thermo Scientific Chemicals
Copper foil, 0.025mm (0.001in) thick, annealed, uncoated, 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper foil, 0.025mm (0.001in) thick, annealed, uncoated, 99.8% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
30 x 1000 cm
30 x 30 cm
30 x 150 cm
Catalog number 046986.RH
also known as 046986-RH
Price (USD)/ Each
490.65
Online exclusive
545.00 
Save 54.35 (10%)
-
Add to cart
Quantity:
30 x 1000 cm
Request bulk or custom format
Price (USD)/ Each
490.65
Online exclusive
545.00 
Save 54.35 (10%)
Add to cart
Copper foil, 0.025mm (0.001in) thick, annealed, uncoated, 99.8% (metals basis), Thermo Scientific Chemicals
Catalog number046986.RH
Price (USD)/ Each
490.65
Online exclusive
545.00 
Save 54.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Copper
FormFoil
Comment0.022 - 0.028mm thick
Total Metal Impurities0.2% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only