Diaion™ PA308(Cl), ion exchange resin, 4% cross-linked, strongly basic porous type I, 1.0 meq/ml on PS-DVB, Thermo Scientific Chemicals
Diaion™ PA308(Cl), ion exchange resin, 4% cross-linked, strongly basic porous type I, 1.0 meq/ml on PS-DVB, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diaion™ PA308(Cl), ion exchange resin, 4% cross-linked, strongly basic porous type I, 1.0 meq/ml on PS-DVB, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 kg
Catalog number 046990.A1
also known as 046990-A1
Price (USD)/ Each
102.00
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
102.00
Add to cart
Diaion™ PA308(Cl), ion exchange resin, 4% cross-linked, strongly basic porous type I, 1.0 meq/ml on PS-DVB, Thermo Scientific Chemicals
Catalog number046990.A1
Price (USD)/ Each
102.00
-
Add to cart
Specifications
Chemical Name or MaterialDiaion™ PA308 (Cl), ion exchange resin
Physical Form300-1180μm Beads
Name Note4% cross-linked, strongly basic porous type I, 1.0meq/mL on PS-DVB
Recommended StorageAmbient temperatures
TSCAYes
View more
Sugar decolorization

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only