Zirconium boride, 99.5% (metals basis excluding Hf), Thermo Scientific Chemicals
Zirconium boride, 99.5% (metals basis excluding Hf), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium boride, 99.5% (metals basis excluding Hf), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 047030.18
Price (USD)/ Each
-
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialZirconium boride
Melting Point3000°C
CAS12045-64-6
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H228
Flammable solid.
View more
Zirconium Boride is used as an aerospace refractory, in cutting tools, and to protect thermocouple tubes. It is also acts as an antioxidant in carbon-bonded refractory ceramics, neutron absorber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only