Thermo Scientific Chemicals

Iron(III) oxide, alpha-phase, nanopowder, 98% (metals basis), Thermo Scientific Chemicals

Catalog number: 047044.22
100 g, Each
Thermo Scientific Chemicals

Iron(III) oxide, alpha-phase, nanopowder, 98% (metals basis), Thermo Scientific Chemicals

Catalog number: 047044.22
100 g, Each
Quantity
Catalog number: 047044.22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1309-37-1
IUPAC Name
trioxodiiron
Molecular Formula
Fe2O3
InChI Key
JEIPFZHSYJVQDO-UHFFFAOYSA-N
SMILES
[Fe][Fe](=O)(=O)=O
Total Metal Impurities
2% max.
Particle Size
30-50 nm

Description

Ferric oxide is used for polishing precious metals and diamonds, as well as in cosmetics. Synthetic iron oxide pigments are one of the most popular color pigments worldwide for coloring concrete. Other uses includes: coating, printing ink, paint, also as coloring agent for building material, rubber, paper-making. In producing ferrites . Also used in recording tapes and pyrotechnics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text