Iron(III) oxide, alpha-phase, nanopowder, 98% (metals basis), Thermo Scientific Chemicals
Iron(III) oxide, alpha-phase, nanopowder, 98% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iron(III) oxide, alpha-phase, nanopowder, 98% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 047044.36
Price (USD)/ Each
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS1309-37-1
IUPAC Nametrioxodiiron
Molecular FormulaFe2O3
InChI KeyJEIPFZHSYJVQDO-UHFFFAOYSA-N
SMILES[Fe][Fe](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities2% max.
Particle Size30-50 nm
Ferric oxide is used for polishing precious metals and diamonds, as well as in cosmetics. Synthetic iron oxide pigments are one of the most popular color pigments worldwide for coloring concrete. Other uses includes: coating, printing ink, paint, also as coloring agent for building material, rubber, paper-making. In producing ferrites . Also used in recording tapes and pyrotechnics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only