Thermo Scientific™

Manganese(III) acetate dihydrate, 97%, Thermo Scientific Chemicals

Catalog number: 047050.09
10 g, Each
Thermo Scientific™

Manganese(III) acetate dihydrate, 97%, Thermo Scientific Chemicals

Catalog number: 047050.09
10 g, Each
Quantity
Catalog number: 047050.09
also known as 047050-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19513-05-4
IUPAC Name
manganese(3+) triacetate
Molecular Formula
C6H9MnO6
InChI Key
AHSBSUVHXDIAEY-UHFFFAOYSA-K
SMILES
[Mn+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Assay (unspecified)
96.1% min

Description

Manganese(III) acetate dihydrate, is used as a mild and selective oxidizing agent. It is also a reagent used for radical cyclizations and α-keto-acetoxylation. Mangese triacetate has been used as a single electron oxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text