Chemical Identifiers
CAS862121-57-1
IUPAC Namestrontium(2+) bis(trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane)
Molecular FormulaC4F12N2O8S4Sr
InChI KeyDIMYICIXZTVMOY-UHFFFAOYSA-N
SMILES[Sr++].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
View more