Nickel bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals
Nickel bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
5 g
Catalog number 047102.03
also known as 047102-03
Price (USD)/ Each
117.65
Online exclusive
130.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
117.65
Online exclusive
130.00 
Save 12.35 (10%)
Add to cart
Nickel bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals
Catalog number047102.03
Price (USD)/ Each
117.65
Online exclusive
130.00 
Save 12.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialNickel bis(trifluoromethylsulfonyl)imide
CAS207861-63-0
ColorGreen
Health Hazard 1H314-H317-H335-H350
Health Hazard 2GHS H Statement
H350-H314-H318-H317
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
View more
Nickel(II) Bis(trifluoromethanesulfonyl)imide is used as Surface treatment, Synthesis and catalysis. It is also used as a ionic liquid in synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only