n-Hexane, anhydrous, over molecular sieves, packaged under argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
n-Hexane, anhydrous, over molecular sieves, packaged under argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Hexane, anhydrous, over molecular sieves, packaged under argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Catalog number 047104.AK
also known as 047104-AK
Price (USD)/ Each
39.65
Online exclusive
44.30 
Save 4.65 (11%)
-
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)/ Each
39.65
Online exclusive
44.30 
Save 4.65 (11%)
Add to cart
n-Hexane, anhydrous, over molecular sieves, packaged under argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Catalog number047104.AK
Price (USD)/ Each
39.65
Online exclusive
44.30 
Save 4.65 (11%)
-
Add to cart
Chemical Identifiers
CAS110-54-3
IUPAC Namehexane
Molecular FormulaC6H14
InChI KeyVLKZOEOYAKHREP-UHFFFAOYSA-N
SMILESCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Residue on Evaporation≤5 ppm
Assay from Supplier's CofA≥95.0% (as n-Hexane)
Assay from Supplier's CofA≥99.0% (as C6 Hydrocarbons)
Impurity contentSulfur compounds (as sulfur): ≤0.005%
View more
n-Hexane is used as a solvent, paint thinner, and chemical reaction medium. It is used to extract edible oils from seeds and vegetables, as a special-use solvent, and as a cleaning agent. Hexanes are commonly used in chromatography as a non-polar solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only