Thermo Scientific Chemicals

n-Hexane, anhydrous, over molecular sieves, packaged under argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 047104.K2
1 L, Each
Thermo Scientific Chemicals

n-Hexane, anhydrous, over molecular sieves, packaged under argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 047104.K2
1 L, Each
Quantity
Catalog number: 047104.K2
also known as 047104-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-54-3
IUPAC Name
hexane
Molecular Formula
C6H14
InChI Key
VLKZOEOYAKHREP-UHFFFAOYSA-N
SMILES
CCCCCC
Form
Liquid
Residue on Evaporation
≤5 ppm
Assay from Supplier's CofA
≥95.0% (as n-Hexane)
Assay from Supplier's CofA
≥99.0% (as C6 Hydrocarbons)
Impurity content
Sulfur compounds (as sulfur): ≤0.005%

Description

n-Hexane is used as a solvent, paint thinner, and chemical reaction medium. It is used to extract edible oils from seeds and vegetables, as a special-use solvent, and as a cleaning agent. Hexanes are commonly used in chromatography as a non-polar solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text