Thermo Scientific Chemicals

Indium(III) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 047108.04
2 g, Each
Thermo Scientific Chemicals

Indium(III) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 047108.04
2 g, Each
Quantity
Catalog number: 047108.04
also known as 047108-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13510-35-5
IUPAC Name
indium(3+) triiodide
Molecular Formula
I3In
InChI Key
RMUKCGUDVKEQPL-UHFFFAOYSA-K
SMILES
[In+3].[I-].[I-].[I-]
Appearance (Color)
White
Form
Beads
Total Metal Impurities
0.001% max.
Particle Size
-10 Mesh

Description

It is used in the preparation vinyl and methyl indates which undergo regioselective reactions with cinnamyl bromide. It is used as an iodide getter in the Cativa Process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text