Thermo Scientific Chemicals

Potassium bromide, Premium FTIR Grade, Thermo Scientific Chemicals

Catalog number: 047111.KN
100 Pc., Each
Thermo Scientific Chemicals

Potassium bromide, Premium FTIR Grade, Thermo Scientific Chemicals

Catalog number: 047111.KN
100 Pc., Each
Quantity
Catalog number: 047111.KN
also known as 047111-KN
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7758-02-3
IUPAC Name
potassium bromide
Molecular Formula
BrK
InChI Key
IOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES
[K+].[Br-]
Form
Powder
Refractive Index
1.559

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text