n-Pentane, anhydrous, 99.8+%, over molecular sieves, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
n-Pentane, anhydrous, 99.8+%, over molecular sieves, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Pentane, anhydrous, 99.8+%, over molecular sieves, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Catalog number 047115.AK
also known as 047115-AK
Price (USD)/ Each
30.65
Online exclusive
33.90 
Save 3.25 (10%)
-
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)/ Each
30.65
Online exclusive
33.90 
Save 3.25 (10%)
Add to cart
n-Pentane, anhydrous, 99.8+%, over molecular sieves, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Catalog number047115.AK
Price (USD)/ Each
30.65
Online exclusive
33.90 
Save 3.25 (10%)
-
Add to cart
Chemical Identifiers
CAS109-66-0
IUPAC Namepentane
Molecular FormulaC5H12
InChI KeyOFBQJSOFQDEBGM-UHFFFAOYSA-N
SMILESCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay from Supplier's CofA≥99.5% (GC)
Residual water≤0.02%
FormLiqiud
Residue on Evaporation≤5 ppm
n-Pentane is used in consumer merchandise such as spot lifters/cleaners and foaming shave gels and in commercial products such as a blowing agent for expanded polystyrene (foam) insulation. It may also be used as a solvent (in pure form) in laboratory, chemical analysis, and other settings where a highly volatile non-polar solvent is needed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only